Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611471 MIM: 602597 | ||||||||||||||||||||
Literature Links: |
ACP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACP6 - acid phosphatase 6, lysophosphatidic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323625.1 | Intron | NP_001310554.1 | ||||
NM_016361.4 | Intron | NP_057445.4 | ||||
XM_011509601.2 | Intron | XP_011507903.1 | ||||
XM_017001422.1 | Intron | XP_016856911.1 |
BCL9 - B-cell CLL/lymphoma 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |