Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615456 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMOD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELMOD1 - ELMO domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130037.1 | Intron | NP_001123509.1 | ||||
NM_001308018.1 | Intron | NP_001294947.1 | ||||
NM_018712.3 | Intron | NP_061182.3 | ||||
XM_017017994.1 | Intron | XP_016873483.1 | ||||
XM_017017995.1 | Intron | XP_016873484.1 |
LOC643923 - uncharacterized LOC643923 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |