Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 103850 MIM: 602035 | ||||||||||||||||||||
Literature Links: |
ALDOA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDOA - aldolase, fructose-bisphosphate A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000034.3 | Intron | NP_000025.1 | ||||
NM_001127617.2 | Intron | NP_001121089.1 | ||||
NM_001243177.1 | Intron | NP_001230106.1 | ||||
NM_184041.2 | Intron | NP_908930.1 | ||||
NM_184043.2 | Intron | NP_908932.1 | ||||
XM_011545768.2 | Intron | XP_011544070.1 |
PPP4C - protein phosphatase 4 catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |