Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4B1 - cytochrome P450 family 4 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000779.3 | Intron | NP_000770.2 | ||||
NM_001099772.1 | Intron | NP_001093242.1 | ||||
NM_001319161.1 | Intron | NP_001306090.1 | ||||
NM_001319162.1 | Intron | NP_001306091.1 | ||||
NM_001319163.1 | Intron | NP_001306092.1 | ||||
XM_011540832.1 | Intron | XP_011539134.1 | ||||
XM_011540833.1 | Intron | XP_011539135.1 | ||||
XM_017000466.1 | Intron | XP_016855955.1 |