Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609744 MIM: 173391 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CADM4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CADM4 - cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAUR - plasminogen activator, urokinase receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005376.2 | 1340 | Intron | NP_001005376.1 | |||
NM_001005377.2 | 1340 | UTR 3 | NP_001005377.1 | |||
NM_001301037.1 | 1340 | UTR 3 | NP_001287966.1 | |||
NM_002659.3 | 1340 | UTR 3 | NP_002650.1 | |||
XM_005258990.4 | 1340 | Intron | XP_005259047.1 | |||
XM_011527027.2 | 1340 | UTR 3 | XP_011525329.1 | |||
XM_011527028.2 | 1340 | UTR 3 | XP_011525330.1 | |||
XM_011527029.1 | 1340 | Intron | XP_011525331.1 | |||
XM_011527030.2 | 1340 | UTR 3 | XP_011525332.1 | |||
XM_011527031.2 | 1340 | UTR 3 | XP_011525333.1 | |||
XM_017026872.1 | 1340 | UTR 3 | XP_016882361.1 | |||
XM_017026873.1 | 1340 | UTR 3 | XP_016882362.1 |
Set Membership: |
HapMap |