Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAUR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLAUR - plasminogen activator, urokinase receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005376.2 | 801 | Silent Mutation | CAC,CAT | H,H 251 | NP_001005376.1 | |
NM_001005377.2 | 801 | Silent Mutation | CAC,CAT | H,H 206 | NP_001005377.1 | |
NM_001301037.1 | 801 | Intron | NP_001287966.1 | |||
NM_002659.3 | 801 | Silent Mutation | CAC,CAT | H,H 251 | NP_002650.1 | |
XM_005258990.4 | 801 | Silent Mutation | CAC,CAT | H,H 251 | XP_005259047.1 | |
XM_011527027.2 | 801 | Silent Mutation | CAC,CAT | H,H 203 | XP_011525329.1 | |
XM_011527028.2 | 801 | UTR 3 | XP_011525330.1 | |||
XM_011527029.1 | 801 | UTR 3 | XP_011525331.1 | |||
XM_011527030.2 | 801 | Intron | XP_011525332.1 | |||
XM_011527031.2 | 801 | Intron | XP_011525333.1 | |||
XM_017026872.1 | 801 | Silent Mutation | CAC,CAT | H,H 246 | XP_016882361.1 | |
XM_017026873.1 | 801 | Silent Mutation | CAC,CAT | H,H 127 | XP_016882362.1 |