Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605252 | ||||||||||||||||||||
Literature Links: |
POLI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLI - DNA polymerase iota | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007195.2 | Intron | NP_009126.2 | ||||
XM_005258188.3 | Intron | XP_005258245.1 | ||||
XM_005258192.4 | Intron | XP_005258249.1 | ||||
XM_006722378.2 | Intron | XP_006722441.1 | ||||
XM_011525796.2 | Intron | XP_011524098.1 | ||||
XM_011525797.1 | Intron | XP_011524099.1 |