Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604699 MIM: 608833 MIM: 603361 | ||||||||||||||||||||
Literature Links: |
ARFRP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARFRP1 - ADP ribosylation factor related protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134758.3 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_001128230.1 | |
NM_001267544.2 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_001254473.1 | |
NM_001267545.2 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_001254474.1 | |
NM_001267546.2 | 467 | Silent Mutation | AGG,CGG | R,R 60 | NP_001254475.1 | |
NM_001267547.2 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_001254476.1 | |
NM_001267548.2 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_001254477.1 | |
NM_001267549.2 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_001254478.1 | |
NM_003224.5 | 467 | Silent Mutation | AGG,CGG | R,R 107 | NP_003215.1 | |
XM_011528482.2 | 467 | Silent Mutation | AGG,CGG | R,R 107 | XP_011526784.1 | |
XM_011528483.1 | 467 | Silent Mutation | AGG,CGG | R,R 107 | XP_011526785.1 | |
XM_017027576.1 | 467 | Silent Mutation | AGG,CGG | R,R 107 | XP_016883065.1 |
RTEL1 - regulator of telomere elongation helicase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTEL1-TNFRSF6B - RTEL1-TNFRSF6B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF6B - TNF receptor superfamily member 6b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZGPAT - zinc finger CCCH-type and G-patch domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |