Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CES4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CES4A - carboxylesterase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190201.1 | 1285 | Silent Mutation | GGG,GGT | G,G 226 | NP_001177130.1 | |
NM_001190202.1 | 1285 | Silent Mutation | GGG,GGT | G,G 230 | NP_001177131.1 | |
NM_001318506.1 | 1285 | Silent Mutation | GGG,GGT | G,G 130 | NP_001305435.1 | |
NM_173815.6 | 1285 | Silent Mutation | GGG,GGT | G,G 324 | NP_776176.5 | |
XM_005255896.4 | 1285 | Silent Mutation | GGG,GGT | G,G 347 | XP_005255953.1 | |
XM_011523021.2 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_011521323.1 | |
XM_011523023.2 | 1285 | Silent Mutation | GGG,GGT | G,G 387 | XP_011521325.1 | |
XM_011523024.2 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_011521326.1 | |
XM_011523025.2 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_011521327.1 | |
XM_011523026.2 | 1285 | Silent Mutation | GGG,GGT | G,G 351 | XP_011521328.1 | |
XM_011523027.2 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_011521329.1 | |
XM_011523028.2 | 1285 | Silent Mutation | GGG,GGT | G,G 339 | XP_011521330.1 | |
XM_011523029.2 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_011521331.1 | |
XM_011523030.2 | 1285 | Silent Mutation | GGG,GGT | G,G 347 | XP_011521332.1 | |
XM_017023160.1 | 1285 | Silent Mutation | GGG,GGT | G,G 387 | XP_016878649.1 | |
XM_017023161.1 | 1285 | Silent Mutation | GGG,GGT | G,G 387 | XP_016878650.1 | |
XM_017023162.1 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_016878651.1 | |
XM_017023163.1 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_016878652.1 | |
XM_017023164.1 | 1285 | Silent Mutation | GGG,GGT | G,G 387 | XP_016878653.1 | |
XM_017023165.1 | 1285 | Silent Mutation | GGG,GGT | G,G 387 | XP_016878654.1 | |
XM_017023166.1 | 1285 | Silent Mutation | GGG,GGT | G,G 391 | XP_016878655.1 | |
XM_017023167.1 | 1285 | Silent Mutation | GGG,GGT | G,G 226 | XP_016878656.1 | |
XM_017023168.1 | 1285 | Silent Mutation | GGG,GGT | G,G 226 | XP_016878657.1 |