Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC36 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRRC36 - leucine rich repeat containing 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161575.1 | Intron | NP_001155047.1 | ||||
NM_018296.5 | Intron | NP_060766.5 | ||||
XM_005256025.2 | Intron | XP_005256082.1 | ||||
XM_005256026.2 | Intron | XP_005256083.1 | ||||
XM_005256027.2 | Intron | XP_005256084.1 | ||||
XM_005256028.1 | Intron | XP_005256085.1 | ||||
XM_011523200.1 | Intron | XP_011521502.1 | ||||
XM_011523201.1 | Intron | XP_011521503.1 | ||||
XM_011523202.2 | Intron | XP_011521504.1 | ||||
XM_011523203.1 | Intron | XP_011521505.1 | ||||
XM_011523204.1 | Intron | XP_011521506.1 | ||||
XM_011523205.1 | Intron | XP_011521507.1 | ||||
XM_011523206.2 | Intron | XP_011521508.1 | ||||
XM_017023400.1 | Intron | XP_016878889.1 | ||||
XM_017023401.1 | Intron | XP_016878890.1 | ||||
XM_017023402.1 | Intron | XP_016878891.1 |