Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176941 | ||||||||||||||||||||
Literature Links: |
TYK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TYK2 - tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003331.4 | 3164 | Missense Mutation | CAC,CAG | H,Q 1017 | NP_003322.3 | |
XM_011528245.1 | 3164 | Missense Mutation | CAC,CAG | H,Q 1017 | XP_011526547.1 | |
XM_011528246.2 | 3164 | Missense Mutation | CAC,CAG | H,Q 918 | XP_011526548.1 | |
XM_011528247.1 | 3164 | Missense Mutation | CAC,CAG | H,Q 918 | XP_011526549.1 | |
XM_011528249.2 | 3164 | Missense Mutation | CAC,CAG | H,Q 575 | XP_011526551.1 | |
XM_011528251.1 | 3164 | Missense Mutation | CAC,CAG | H,Q 436 | XP_011526553.1 | |
XM_011528252.1 | 3164 | Intron | XP_011526554.1 |