Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611509 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPN - centromere protein N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100624.2 | Intron | NP_001094094.2 | ||||
NM_001100625.2 | Intron | NP_001094095.2 | ||||
NM_001270473.1 | Intron | NP_001257402.1 | ||||
NM_001270474.1 | Intron | NP_001257403.1 | ||||
NM_018455.5 | Intron | NP_060925.2 | ||||
XM_006721236.3 | Intron | XP_006721299.1 | ||||
XM_017023456.1 | Intron | XP_016878945.1 |
CMC2 - C-X9-C motif containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |