Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610514 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JADE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
JADE1 - jade family PHD finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287437.1 | Intron | NP_001274366.1 | ||||
NM_001287439.1 | Intron | NP_001274368.1 | ||||
NM_001287440.1 | Intron | NP_001274369.1 | ||||
NM_001287441.1 | Intron | NP_001274370.1 | ||||
NM_001287442.1 | Intron | NP_001274371.1 | ||||
NM_001287443.1 | Intron | NP_001274372.1 | ||||
NM_024900.4 | Intron | NP_079176.2 | ||||
NM_199320.3 | Intron | NP_955352.1 | ||||
XM_005263232.1 | Intron | XP_005263289.1 | ||||
XM_017008626.1 | Intron | XP_016864115.1 | ||||
XM_017008627.1 | Intron | XP_016864116.1 |
Set Membership: |
HapMap |