Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGACTCAGGTTGTGGCGGATGG[T/C]GTTCTGTGGAAGGCCGGGGACAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300859 MIM: 300292 | ||||||||||||||||||||
Literature Links: |
CCDC22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC22 - coiled-coil domain containing 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014008.4 | 1338 | Intron | NP_054727.1 | |||
XM_005272599.3 | 1338 | Intron | XP_005272656.1 |
FOXP3 - forkhead box P3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114377.1 | 1338 | Missense Mutation | ACC,GCC | T,A 349 | NP_001107849.1 | |
NM_014009.3 | 1338 | Missense Mutation | ACC,GCC | T,A 384 | NP_054728.2 | |
XM_006724533.2 | 1338 | Missense Mutation | ACC,GCC | T,A 407 | XP_006724596.2 | |
XM_011543916.2 | 1338 | Missense Mutation | ACC,GCC | T,A 517 | XP_011542218.1 | |
XM_017029565.1 | 1338 | Missense Mutation | ACC,GCC | T,A 469 | XP_016885054.1 | |
XM_017029566.1 | 1338 | Missense Mutation | ACC,GCC | T,A 457 | XP_016885055.1 | |
XM_017029567.1 | 1338 | Missense Mutation | ACC,GCC | T,A 426 | XP_016885056.1 |