Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCAGTAGTCTCTGGAGAGGCT[C/G]TGCACATCCTCTCTTTGAACAGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KBTBD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KBTBD11 - kelch repeat and BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014867.2 | Intron | NP_055682.1 | ||||
XM_011534771.2 | Intron | XP_011533073.1 | ||||
XM_011534772.2 | Intron | XP_011533074.1 | ||||
XM_017014114.1 | Intron | XP_016869603.1 | ||||
XM_017014115.1 | Intron | XP_016869604.1 | ||||
XM_017014116.1 | Intron | XP_016869605.1 | ||||
XM_017014117.1 | Intron | XP_016869606.1 |
KBTBD11-OT1 - KBTBD11 overlapping transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928058 - uncharacterized LOC101928058 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |