Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605451 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAK4 - p21 (RAC1) activated kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014831.2 | 102 | UTR 5 | NP_001014831.1 | |||
NM_001014832.1 | 102 | UTR 5 | NP_001014832.1 | |||
NM_001014834.2 | 102 | UTR 5 | NP_001014834.1 | |||
NM_001014835.1 | 102 | UTR 5 | NP_001014835.1 | |||
NM_005884.3 | 102 | UTR 5 | NP_005875.1 | |||
XM_011526316.1 | 102 | UTR 5 | XP_011524618.1 | |||
XM_011526317.2 | 102 | Intron | XP_011524619.1 | |||
XM_011526318.2 | 102 | Intron | XP_011524620.1 | |||
XM_011526319.2 | 102 | Intron | XP_011524621.1 | |||
XM_011526320.1 | 102 | UTR 5 | XP_011524622.1 |