Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603558 | ||||||||||||||||||||
Literature Links: |
MTMR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTMR3 - myotubularin related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021090.3 | Intron | NP_066576.1 | ||||
NM_153050.2 | Intron | NP_694690.1 | ||||
NM_153051.2 | Intron | NP_694691.1 | ||||
XM_005261803.2 | Intron | XP_005261860.1 | ||||
XM_005261804.3 | Intron | XP_005261861.1 | ||||
XM_005261805.2 | Intron | XP_005261862.1 | ||||
XM_005261806.2 | Intron | XP_005261863.1 | ||||
XM_005261808.2 | Intron | XP_005261865.1 | ||||
XM_017029028.1 | Intron | XP_016884517.1 | ||||
XM_017029029.1 | Intron | XP_016884518.1 | ||||
XM_017029030.1 | Intron | XP_016884519.1 | ||||
XM_017029031.1 | Intron | XP_016884520.1 | ||||
XM_017029032.1 | Intron | XP_016884521.1 | ||||
XM_017029033.1 | Intron | XP_016884522.1 | ||||
XM_017029034.1 | Intron | XP_016884523.1 |