Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 167415 | ||||||||||||||||||||
Literature Links: |
PAX8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAX8 - paired box 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003466.3 | 1506 | Missense Mutation | CGC,GGC | R,G 418 | NP_003457.1 | |
NM_013952.3 | 1506 | Silent Mutation | GGC,GGG | G,G 391 | NP_039246.1 | |
NM_013953.3 | 1506 | Silent Mutation | GGC,GGG | G,G 314 | NP_039247.1 | |
NM_013992.3 | 1506 | Silent Mutation | GGC,GGG | G,G 280 | NP_054698.1 | |
XM_011511790.1 | 1506 | Missense Mutation | CGC,GGC | R,G 475 | XP_011510092.1 | |
XM_011511791.1 | 1506 | Silent Mutation | GGC,GGG | G,G 493 | XP_011510093.1 | |
XM_011511792.1 | 1506 | Silent Mutation | GGC,GGG | G,G 448 | XP_011510094.1 | |
XM_011511793.1 | 1506 | Missense Mutation | CGC,GGC | R,G 412 | XP_011510095.1 | |
XM_011511794.1 | 1506 | Missense Mutation | CGC,GGC | R,G 355 | XP_011510096.1 | |
XM_017004878.1 | 1506 | Intron | XP_016860367.1 | |||
XM_017004879.1 | 1506 | Intron | XP_016860368.1 |