Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608377 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARFGAP1 - ADP ribosylation factor GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281482.1 | 65 | Intron | NP_001268411.1 | |||
NM_001281483.1 | 65 | Intron | NP_001268412.1 | |||
NM_001281484.1 | 65 | Intron | NP_001268413.1 | |||
NM_018209.3 | 65 | Intron | NP_060679.1 | |||
NM_175609.2 | 65 | Intron | NP_783202.1 | |||
XM_006723823.2 | 65 | Intron | XP_006723886.1 | |||
XM_011528901.1 | 65 | Intron | XP_011527203.1 | |||
XM_017027950.1 | 65 | UTR 5 | XP_016883439.1 |
Set Membership: |
HapMap |