Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189971 MIM: 612478 | ||||||||||||||||||||
Literature Links: |
ACTL10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTL10 - actin like 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
E2F1 - E2F transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005225.2 | 1072 | Missense Mutation | AAC,ACC | N,T 311 | NP_005216.1 |
NECAB3 - N-terminal EF-hand calcium binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |