Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTCATTACTCTTATGTAGGTAA[C/T]ACCACAGTGTCTAAAGGGCTCGCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610343 MIM: 608879 | ||||||||||||||||||||
Literature Links: |
C2CD4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2CD4A - C2 calcium dependent domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207322.2 | 2288 | UTR 3 | NP_997205.2 |
LOC101928907 - uncharacterized LOC101928907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS13C - vacuolar protein sorting 13 homolog C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |