Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176310 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PBX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PBX1 - PBX homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204961.1 | Intron | NP_001191890.1 | ||||
NM_001204963.1 | Intron | NP_001191892.1 | ||||
NM_002585.3 | Intron | NP_002576.1 | ||||
XM_005245228.3 | Intron | XP_005245285.1 | ||||
XM_005245229.3 | Intron | XP_005245286.1 | ||||
XM_011509590.1 | Intron | XP_011507892.1 | ||||
XM_011509591.2 | Intron | XP_011507893.1 | ||||
XM_011509592.1 | Intron | XP_011507894.1 | ||||
XM_017001395.1 | Intron | XP_016856884.1 | ||||
XM_017001396.1 | Intron | XP_016856885.1 |
Set Membership: |
HapMap |