Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTTGCTGCCATGGCAATGCTGC[C/G]TAACAACTTAAGAGAAACGTGAAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606504 MIM: 614801 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASC3 - cancer susceptibility candidate 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSL1 - male specific lethal 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012241.1 | Intron | NP_001012241.1 | ||||
XM_005257298.4 | Intron | XP_005257355.1 | ||||
XM_005257299.4 | Intron | XP_005257356.3 |
Set Membership: |
HapMap |