Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCCGGCCGAGATGGCGGATCCA[A/G]CCGAATGCAGCATCAAAGTGATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604593 | ||||||||||||||||||||
Literature Links: |
KIF5C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF5C - kinesin family member 5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004522.2 | 408 | Missense Mutation | ACC,GCC | T,A 5 | NP_004513.1 | |
XM_011511157.2 | 408 | Intron | XP_011509459.1 | |||
XM_017004062.1 | 408 | Missense Mutation | ACC,GCC | T,A 5 | XP_016859551.1 |
LOC101928553 - uncharacterized LOC101928553 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |