Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCTCTGGGACAGACCAACTCAG[A/G]GAGTGGAGGCCCACGCTCTGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606073 MIM: 616499 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDOR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NDOR1 - NADPH dependent diflavin oxidoreductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144026.2 | Intron | NP_001137498.1 | ||||
NM_001144027.2 | Intron | NP_001137499.1 | ||||
NM_001144028.2 | Intron | NP_001137500.1 | ||||
NM_014434.3 | Intron | NP_055249.1 | ||||
XM_011518545.1 | Intron | XP_011516847.1 | ||||
XM_011518547.1 | Intron | XP_011516849.1 | ||||
XM_011518548.2 | Intron | XP_011516850.1 | ||||
XM_017014626.1 | Intron | XP_016870115.1 | ||||
XM_017014627.1 | Intron | XP_016870116.1 |
RNF208 - ring finger protein 208 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM203 - transmembrane protein 203 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |