Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCTCTTCTTCTAAATAGATGG[A/C]AGTCCACAGGCCCCTACTGGATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612405 MIM: 602122 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARL9 - ADP ribosylation factor like GTPase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206919.1 | Intron | NP_996802.1 | ||||
XM_005265719.3 | Intron | XP_005265776.1 | ||||
XM_005265720.3 | Intron | XP_005265777.1 | ||||
XM_006714036.3 | Intron | XP_006714099.1 |
SRP72 - signal recognition particle 72 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |