Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATGTCACTCGGCCAGAGCACTT[C/T]GACTTTGTAAAGCCTGAGGACCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607611 MIM: 608076 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLSCR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLSCR3 - phospholipid scramblase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM256-PLSCR3 - TMEM256-PLSCR3 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNK1 - tyrosine kinase non receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251902.1 | 346 | Silent Mutation | TTC,TTT | F,F 38 | NP_001238831.1 | |
NM_003985.4 | 346 | Silent Mutation | TTC,TTT | F,F 38 | NP_003976.2 | |
XM_011524045.2 | 346 | Silent Mutation | TTC,TTT | F,F 38 | XP_011522347.1 | |
XM_011524046.2 | 346 | Silent Mutation | TTC,TTT | F,F 38 | XP_011522348.1 | |
XM_011524047.2 | 346 | Silent Mutation | TTC,TTT | F,F 38 | XP_011522349.1 | |
XM_011524048.1 | 346 | UTR 5 | XP_011522350.1 |