Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTAAATGCAAAATATGTGGTAC[A/G]GATATTAAGTGCTGTAGTAGGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614184 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIS3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIS3L2 - DIS3 like 3'-5' exoribonuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257281.1 | Intron | NP_001244210.1 | ||||
NM_001257282.1 | Intron | NP_001244211.1 | ||||
NM_152383.4 | Intron | NP_689596.4 |
Set Membership: |
HapMap |