Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGAGACTCATTTAATTGTACA[A/T]CTTGAACTGATCCTTTTCTGGGGAA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 614184 | |||||||||||||||||||||||||||||
Literature Links: |
DIS3L2 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DIS3L2 - DIS3 like 3'-5' exoribonuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257281.1 | Intron | NP_001244210.1 | ||||
NM_001257282.1 | Intron | NP_001244211.1 | ||||
NM_152383.4 | Intron | NP_689596.4 |
Set Membership: |
HapMap |