Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGAGACCACCTGGCTGGGAGAAG[G/T]CGGAGGAGGCGATGGCTACTATCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603884 MIM: 607281 | ||||||||||||||||||||
Literature Links: |
BAG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAG4 - BCL2 associated athanogene 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204878.1 | 479 | Missense Mutation | GGC,GTC | G,V 66 | NP_001191807.1 | |
NM_004874.3 | 479 | Missense Mutation | GGC,GTC | G,V 66 | NP_004865.1 |
LSM1 - LSM1 homolog, mRNA degradation associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014462.2 | 479 | Intron | NP_055277.1 |