Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGGGGTTGTACTGTAGACAGG[A/T]ATTGGCAGGAAGGGCTGCCTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LONRF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LONRF3 - LON peptidase N-terminal domain and ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031855.2 | Intron | NP_001027026.1 | ||||
NM_001289109.1 | Intron | NP_001276038.1 | ||||
NM_024778.5 | Intron | NP_079054.3 | ||||
XM_005262476.2 | Intron | XP_005262533.1 | ||||
XM_011531398.2 | Intron | XP_011529700.1 | ||||
XM_017029845.1 | Intron | XP_016885334.1 |