Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTAGGATGAAGAGAGAGTCCT[C/T]GGAAGGTGTCATCAGGTATTGACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSTL5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FSTL5 - follistatin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128427.2 | 2876 | Missense Mutation | AAG,GAG | K,E 814 | NP_001121899.1 | |
NM_001128428.2 | 2876 | Missense Mutation | AAG,GAG | K,E 805 | NP_001121900.1 | |
NM_020116.4 | 2876 | Missense Mutation | AAG,GAG | K,E 815 | NP_064501.2 | |
XM_011532126.1 | 2876 | Missense Mutation | AAG,GAG | K,E 806 | XP_011530428.1 |
Set Membership: |
HapMap |