Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACATACTTTTGATGTTTGGGTC[C/T]GGCTTTTTTACTGACGTGCCTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611406 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNC1LI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYNC1LI2 - dynein cytoplasmic 1 light intermediate chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286157.1 | 1276 | Silent Mutation | CCA,CCG | P,P 338 | NP_001273086.1 | |
NM_001323955.1 | 1276 | Silent Mutation | CCA,CCG | P,P 376 | NP_001310884.1 | |
NM_006141.2 | 1276 | Silent Mutation | CCA,CCG | P,P 415 | NP_006132.1 | |
XM_017023008.1 | 1276 | Silent Mutation | CCA,CCG | P,P 415 | XP_016878497.1 |