Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTAGCAAAAGGCCATTCGCTC[C/T]TTCATTCATCATAATGGCAGCAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611353 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INTS10 - integrator complex subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018142.2 | Intron | NP_060612.2 | ||||
XM_005273555.3 | Intron | XP_005273612.1 | ||||
XM_005273556.3 | Intron | XP_005273613.1 | ||||
XM_005273557.3 | Intron | XP_005273614.1 | ||||
XM_006716355.2 | Intron | XP_006716418.1 | ||||
XM_017013606.1 | Intron | XP_016869095.1 | ||||
XM_017013607.1 | Intron | XP_016869096.1 | ||||
XM_017013608.1 | Intron | XP_016869097.1 | ||||
XM_017013609.1 | Intron | XP_016869098.1 |