Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604891 | ||||||||||||||||||||
Literature Links: |
NCKAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCKAP1 - NCK associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013436.4 | 3531 | Missense Mutation | TCT,TTT | S,F 1095 | NP_038464.1 | |
NM_205842.2 | 3531 | Missense Mutation | TCT,TTT | S,F 1101 | NP_995314.1 | |
XM_006712200.3 | 3531 | Missense Mutation | TCT,TTT | S,F 1099 | XP_006712263.1 | |
XM_006712201.3 | 3531 | Missense Mutation | TCT,TTT | S,F 1093 | XP_006712264.1 |