Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTTCTGCCCATCCTTTGCACA[C/G]AATATGGAAATATAGGAGGTATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600697 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBBP5 - RB binding protein 5, histone lysine methyltransferase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193272.1 | 2271 | UTR 3 | NP_001180201.1 | |||
NM_001193273.1 | 2271 | UTR 3 | NP_001180202.1 | |||
NM_005057.3 | 2271 | UTR 3 | NP_005048.2 | |||
XM_011509833.2 | 2271 | UTR 3 | XP_011508135.1 |
TMEM81 - transmembrane protein 81 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |