Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAAGCATTAAGCGTTGGCGAGT[A/G]GGAAAAAACACAGAACTGTTTGTCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
KBTBD12 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KBTBD12 - kelch repeat and BTB domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207335.2 | Intron | NP_997218.2 | ||||
XM_005247167.1 | Intron | XP_005247224.1 | ||||
XM_005247168.4 | Intron | XP_005247225.1 | ||||
XM_011512490.2 | Intron | XP_011510792.1 | ||||
XM_011512491.2 | Intron | XP_011510793.1 | ||||
XM_011512492.2 | Intron | XP_011510794.1 |