Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGAAGATCCTGAGTGGCTGGGA[C/G]GTGACTTGCTTCTCCCCAGATGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607465 MIM: 611695 | ||||||||||||||||||||
Literature Links: |
CDAN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDAN1 - codanin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTBK2 - tau tubulin kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173500.3 | 3893 | Missense Mutation | CCT,CGT | P,R 1191 | NP_775771.3 | |
XM_005254171.4 | 3893 | Missense Mutation | CCT,CGT | P,R 1197 | XP_005254228.1 | |
XM_005254173.4 | 3893 | Missense Mutation | CCT,CGT | P,R 1122 | XP_005254230.1 | |
XM_006720402.3 | 3893 | Missense Mutation | CCT,CGT | P,R 1186 | XP_006720465.1 | |
XM_006720403.3 | 3893 | Missense Mutation | CCT,CGT | P,R 1122 | XP_006720466.1 | |
XM_017021950.1 | 3893 | Missense Mutation | CCT,CGT | P,R 1098 | XP_016877439.1 |