Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGAAAGAGTGACATCTGCTGTTT[C/T]TCATGCTCAGCATCCAATGCCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607590 MIM: 123835 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BBS7 - Bardet-Biedl syndrome 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018190.3 | Intron | NP_060660.2 | ||||
NM_176824.2 | Intron | NP_789794.1 | ||||
XM_005263106.3 | Intron | XP_005263163.1 | ||||
XM_011532079.2 | Intron | XP_011530381.1 | ||||
XM_011532080.2 | Intron | XP_011530382.1 | ||||
XM_011532081.2 | Intron | XP_011530383.1 | ||||
XM_017008357.1 | Intron | XP_016863846.1 | ||||
XM_017008358.1 | Intron | XP_016863847.1 |
CCNA2 - cyclin A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |