Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGTCTGTTTCCTCATATGCGA[C/G]GAAACAGGCAGAGAGACTGCAATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EFL1 - elongation factor like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040610.2 | Intron | NP_001035700.1 | ||||
NM_001322844.1 | Intron | NP_001309773.1 | ||||
NM_001322845.1 | Intron | NP_001309774.1 | ||||
NM_024580.5 | Intron | NP_078856.4 | ||||
XM_011522000.1 | Intron | XP_011520302.1 |
Set Membership: |
HapMap |