Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCAAAAAGCAGTAAGAAAGTA[A/G]CTATTGAGAAAGAAGGAGGGCCATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607469 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAAA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NAAA - N-acylethanolamine acid amidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042402.1 | 1669 | Intron | NP_001035861.1 | |||
NM_014435.3 | 1669 | UTR 3 | NP_055250.2 | |||
XM_005262920.3 | 1669 | Intron | XP_005262977.1 | |||
XM_005262921.3 | 1669 | Intron | XP_005262978.1 | |||
XM_005262923.2 | 1669 | UTR 3 | XP_005262980.1 | |||
XM_005262924.2 | 1669 | UTR 3 | XP_005262981.1 | |||
XM_005262925.3 | 1669 | Intron | XP_005262982.1 | |||
XM_006714180.2 | 1669 | Intron | XP_006714243.1 | |||
XM_011531852.2 | 1669 | Intron | XP_011530154.1 | |||
XM_011531854.2 | 1669 | Intron | XP_011530156.1 | |||
XM_017008027.1 | 1669 | Intron | XP_016863516.1 | |||
XM_017008028.1 | 1669 | UTR 3 | XP_016863517.1 | |||
XM_017008029.1 | 1669 | Intron | XP_016863518.1 | |||
XM_017008030.1 | 1669 | UTR 3 | XP_016863519.1 |
Set Membership: |
HapMap |