Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCTGTGTCCCACAAAGCCAA[C/T]TCTACCTGTAATGGGAAAAACAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165390 | ||||||||||||||||||||
Literature Links: |
RHOA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHOA - ras homolog family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313941.1 | 319 | Silent Mutation | GAA,GAG | E,E 54 | NP_001300870.1 | |
NM_001313943.1 | 319 | Silent Mutation | GAA,GAG | E,E 54 | NP_001300872.1 | |
NM_001313944.1 | 319 | Intron | NP_001300873.1 | |||
NM_001313945.1 | 319 | UTR 5 | NP_001300874.1 | |||
NM_001313946.1 | 319 | Intron | NP_001300875.1 | |||
NM_001313947.1 | 319 | Intron | NP_001300876.1 | |||
NM_001664.3 | 319 | Silent Mutation | GAA,GAG | E,E 54 | NP_001655.1 |