Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCATTCAGCCCAGAGTTGTTG[C/T]AGACTCATCTATAGAGCTTTTATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605599 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375839 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105375839 - uncharacterized LOC105375839 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYPLA1 - lysophospholipase I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279356.1 | Intron | NP_001266285.1 | ||||
NM_001279357.1 | Intron | NP_001266286.1 | ||||
NM_001279358.1 | Intron | NP_001266287.1 | ||||
NM_001279359.1 | Intron | NP_001266288.1 | ||||
NM_001279360.1 | Intron | NP_001266289.1 | ||||
NM_006330.3 | Intron | NP_006321.1 | ||||
XM_005251127.4 | Intron | XP_005251184.1 | ||||
XM_017012956.1 | Intron | XP_016868445.1 | ||||
XM_017012957.1 | Intron | XP_016868446.1 | ||||
XM_017012958.1 | Intron | XP_016868447.1 | ||||
XM_017012959.1 | Intron | XP_016868448.1 | ||||
XM_017012960.1 | Intron | XP_016868449.1 |
Set Membership: |
HapMap |