Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGGAAGTCTTGGTGAATGATC[G/T]CCCACTGATTCTGTGTAACTTATAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300230 MIM: 300165 MIM: 300028 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA5B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CA5B - carbonic anhydrase 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INE2 - inactivation escape 2 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZRSR2 - zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005089.3 | Intron | NP_005080.1 | ||||
XM_005274597.2 | Intron | XP_005274654.2 | ||||
XM_011545589.2 | Intron | XP_011543891.2 | ||||
XM_011545590.2 | Intron | XP_011543892.2 | ||||
XM_017029881.1 | Intron | XP_016885370.1 | ||||
XM_017029882.1 | Intron | XP_016885371.1 | ||||
XM_017029883.1 | Intron | XP_016885372.1 |
Set Membership: |
HapMap |