Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTATTCTCAGTCTCACAAGGCT[A/G]TGCTCGCTGTCTTCCTTACAAAGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600433 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CBLN2 - cerebellin 2 precursor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182511.3 | Intron | NP_872317.1 | ||||
XM_006722394.3 | Intron | XP_006722457.1 | ||||
XM_011525824.2 | Intron | XP_011524126.1 | ||||
XM_017025559.1 | Intron | XP_016881048.1 | ||||
XM_017025560.1 | Intron | XP_016881049.1 |