Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCGGAGGAAGAGGAGCTGGTCTG[C/T]TGGCTTTGAAGCACCTGATCACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600433 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CBLN2 - cerebellin 2 precursor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182511.3 | 2873 | UTR 3 | NP_872317.1 | |||
XM_006722394.3 | 2873 | UTR 3 | XP_006722457.1 | |||
XM_011525824.2 | 2873 | UTR 3 | XP_011524126.1 | |||
XM_017025559.1 | 2873 | UTR 3 | XP_016881048.1 | |||
XM_017025560.1 | 2873 | UTR 3 | XP_016881049.1 |