Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGAGAGTTTGTAAAAAGACACA[C/T]TCCTCTTACAAGAGTTCATGCTACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607076 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PI15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PI15 - peptidase inhibitor 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324403.1 | 53 | UTR 5 | NP_001311332.1 | |||
NM_015886.4 | 53 | UTR 5 | NP_056970.1 |
Set Membership: |
HapMap |