Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGGAACATGGACGTTATCAAGG[A/T]CAGCTGCCGGCACTCTTGGTAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613922 MIM: 613924 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIPK - lipase family member K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIPN - lipase family member N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102469.1 | 168 | Intron | NP_001095939.1 | |||
XM_005270049.3 | 168 | UTR 5 | XP_005270106.1 | |||
XM_011540083.2 | 168 | UTR 5 | XP_011538385.1 | |||
XM_011540084.2 | 168 | UTR 5 | XP_011538386.1 | |||
XM_011540085.2 | 168 | UTR 5 | XP_011538387.1 | |||
XM_011540086.2 | 168 | UTR 5 | XP_011538388.1 | |||
XM_017016549.1 | 168 | UTR 5 | XP_016872038.1 |
Set Membership: |
HapMap |