Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAAGTGCTATTGCTAAGATGGA[G/T]TCCAAACTCCTTACTGCATATGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607871 MIM: 600678 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO11 - F-box protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190274.1 | Intron | NP_001177203.1 | ||||
NM_025133.4 | Intron | NP_079409.3 | ||||
XM_005264572.4 | Intron | XP_005264629.1 | ||||
XM_005264573.4 | Intron | XP_005264630.1 | ||||
XM_017005015.1 | Intron | XP_016860504.1 | ||||
XM_017005016.1 | Intron | XP_016860505.1 | ||||
XM_017005017.1 | Intron | XP_016860506.1 |
MSH6 - mutS homolog 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |