Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608733 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC39A10 - solute carrier family 39 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127257.1 | Intron | NP_001120729.1 | ||||
NM_020342.2 | Intron | NP_065075.1 | ||||
XM_005246689.4 | Intron | XP_005246746.2 | ||||
XM_011511504.2 | Intron | XP_011509806.1 | ||||
XM_011511505.2 | Intron | XP_011509807.1 | ||||
XM_011511506.2 | Intron | XP_011509808.1 | ||||
XM_011511507.2 | Intron | XP_011509809.1 | ||||
XM_017004522.1 | Intron | XP_016860011.1 | ||||
XM_017004523.1 | Intron | XP_016860012.1 |
Set Membership: |
HapMap |